Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242854.1 | 2943 | UTR 3 | NP_001229783.1 | ||
NM_001242855.1 | 2943 | UTR 3 | NP_001229784.1 | ||
NM_001242856.1 | 2943 | UTR 3 | NP_001229785.1 | ||
NM_012402.3 | 2943 | UTR 3 | NP_036534.1 | ||
XM_005252840.4 | 2943 | UTR 3 | XP_005252897.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001248006.1 | 2943 | Intron | NP_001234935.1 | ||
NM_001248007.1 | 2943 | Intron | NP_001234936.1 | ||
NM_006458.3 | 2943 | Intron | NP_006449.2 | ||
NM_033278.3 | 2943 | Intron | NP_150594.2 | ||
XM_011519844.1 | 2943 | Intron | XP_011518146.1 | ||
XM_011519845.2 | 2943 | Intron | XP_011518147.1 | ||
XM_017017098.1 | 2943 | Intron | XP_016872587.1 | ||
XM_017017099.1 | 2943 | Intron | XP_016872588.1 |