Product Details
- SNP ID
-
rs36032567
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:26207454 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTCCTCCCTCTTCTCCCAAAA[C/G]TTAAGGCCACTAGCATGATTATAGC
- Phenotype
-
MIM: 604477
MIM: 600124
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CBX3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78244699] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CBX3
- Gene Name
- chromobox 3
- Gene
- HNRNPA2B1
- Gene Name
- heterogeneous nuclear ribonucleoprotein A2/B1
There are no transcripts associated with this gene.
View Full Product Details