Product Details
- SNP ID
-
rs6773558
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:150661071 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTAATAAGAAAGATTTATCCCCC[C/T]GCCCCCACCTTGTTTCCAGCGGGGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH6
PubMed Links
Gene Details
- Gene
- ERICH6
- Gene Name
- glutamate rich 6
View Full Product Details