Product Details

SNP ID
rs35225263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103400289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTTAGCAGTTCTCTGGCTTCTA[A/T]TGACTATAGAGCAATTTCGAATATG
Phenotype
MIM: 610815
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A32 PubMed Links

Gene Details

Gene
SLC25A32
Gene Name
solute carrier family 25 member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030780.4 1468 UTR 3 NP_110407.2
XM_017013877.1 1468 UTR 3 XP_016869366.1

View Full Product Details