Product Details

SNP ID
rs1044280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8731212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTATGAGATTTGCCTTTTCAAG[A/C]TGGGAGTCATCCGCCACTTCATTGT
Phenotype
MIM: 615759
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIDINS220 PubMed Links

Gene Details

Gene
KIDINS220
Gene Name
kinase D-interacting substrate 220kDa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020738.2 5006 Missense Mutation CAG,CAT Q1608H NP_065789.1

View Full Product Details