Product Details

SNP ID
rs1254319
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:60437039 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCATCTTCTCCTTCTGGCTTCTCAA[A/G]TAAGTTTCCTAAGGAAGATAAACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf39 PubMed Links

Gene Details

Gene
C14orf39
Gene Name
chromosome 14 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174978.2 1643 Missense Mutation CTT,TTT L524F NP_777638.2
XM_011536690.2 1643 Missense Mutation CTT,TTT L560F XP_011534992.1
XM_011536703.2 1643 Intron XP_011535005.1
XM_011536704.1 1643 Intron XP_011535006.1
XM_017021247.1 1643 Missense Mutation CTT,TTT L559F XP_016876736.1
XM_017021248.1 1643 Missense Mutation CTT,TTT L495F XP_016876737.1
XM_017021249.1 1643 Missense Mutation CTT,TTT L432F XP_016876738.1
XM_017021250.1 1643 Missense Mutation CTT,TTT L415F XP_016876739.1
XM_017021251.1 1643 Missense Mutation CTT,TTT L415F XP_016876740.1

View Full Product Details