Product Details

SNP ID
rs12497433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133669740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGTGAACTCCCCACTCCCACC[C/G]TAACAAATCACCACGATTGCTGCCA
Phenotype
MIM: 190000 MIM: 607760
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TF PubMed Links

Gene Details

Gene
TF
Gene Name
transferrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001063.3 Intron NP_001054.1
XM_017007089.1 Intron XP_016862578.1
XM_017007090.1 Intron XP_016862579.1
Gene
TOPBP1
Gene Name
topoisomerase (DNA) II binding protein 1
There are no transcripts associated with this gene.

View Full Product Details