Product Details

SNP ID
rs1190583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102143339 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTCACAATCGAGGAAGTTTAGA[A/G]TGGGGTTTCTTGTGTGAGGAGATGT
Phenotype
MIM: 140571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90AA1 PubMed Links

Gene Details

Gene
HSP90AA1
Gene Name
heat shock protein 90 alpha family class A member 1
There are no transcripts associated with this gene.

Gene
WDR20
Gene Name
WD repeat domain 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242414.1 Intron NP_001229343.1
NM_001242415.1 Intron NP_001229344.1
NM_001242416.1 Intron NP_001229345.1
NM_001242417.1 Intron NP_001229346.1
NM_001242418.1 Intron NP_001229347.1
NM_001320130.1 Intron NP_001307059.1
NM_144574.3 Intron NP_653175.2
NM_181291.2 Intron NP_851808.1
NM_181308.2 Intron NP_851825.1
XM_006720308.1 Intron XP_006720371.1
XM_006720309.1 Intron XP_006720372.1
XM_006720310.1 Intron XP_006720373.1
XM_006720311.1 Intron XP_006720374.1
XM_006720313.2 Intron XP_006720376.1
XM_006720318.2 Intron XP_006720381.1
XM_006720320.3 Intron XP_006720383.1
XM_011537335.2 Intron XP_011535637.1
XM_011537336.2 Intron XP_011535638.1
XM_011537337.2 Intron XP_011535639.1
XM_011537338.1 Intron XP_011535640.1
XM_011537339.2 Intron XP_011535641.1
XM_011537340.2 Intron XP_011535642.1
XM_011537341.2 Intron XP_011535643.1
XM_011537343.1 Intron XP_011535645.1
XM_011537344.2 Intron XP_011535646.1
XM_011537345.1 Intron XP_011535647.1
XM_011537346.1 Intron XP_011535648.1
XM_011537347.1 Intron XP_011535649.1
XM_011537352.2 Intron XP_011535654.1
XM_011537353.2 Intron XP_011535655.1
XM_011537355.2 Intron XP_011535657.1
XM_011537356.2 Intron XP_011535658.1
XM_011537358.2 Intron XP_011535660.1
XM_017021766.1 Intron XP_016877255.1
XM_017021767.1 Intron XP_016877256.1
XM_017021768.1 Intron XP_016877257.1
XM_017021770.1 Intron XP_016877259.1
XM_017021771.1 Intron XP_016877260.1
XM_017021772.1 Intron XP_016877261.1
XM_017021773.1 Intron XP_016877262.1
XM_017021774.1 Intron XP_016877263.1
XM_017021775.1 Intron XP_016877264.1
XM_017021776.1 Intron XP_016877265.1
XM_017021777.1 Intron XP_016877266.1
XM_017021778.1 Intron XP_016877267.1
XM_017021779.1 Intron XP_016877268.1
XM_017021780.1 Intron XP_016877269.1
XM_017021781.1 Intron XP_016877270.1
XM_017021782.1 Intron XP_016877271.1
XM_017021783.1 Intron XP_016877272.1
XM_017021784.1 Intron XP_016877273.1
XM_017021785.1 Intron XP_016877274.1

View Full Product Details