Product Details

SNP ID
rs6672194
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:67092283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAATGCAACTTGTTTCTAAGT[G/T]GAATTCTGCCTTTAAACTGTTTGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf141 PubMed Links

Gene Details

Gene
C1orf141
Gene Name
chromosome 1 open reading frame 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276351.1 2158 UTR 3 NP_001263280.1
NM_001276352.1 2158 UTR 3 NP_001263281.1
XM_011541465.2 2158 UTR 3 XP_011539767.1
XM_011541466.2 2158 UTR 3 XP_011539768.1
XM_011541467.1 2158 UTR 3 XP_011539769.1
XM_011541469.1 2158 UTR 3 XP_011539771.1
XM_011541472.1 2158 Intron XP_011539774.1
XM_011541473.2 2158 Intron XP_011539775.1
XM_011541475.1 2158 Intron XP_011539777.1
XM_017001276.1 2158 UTR 3 XP_016856765.1
XM_017001277.1 2158 Intron XP_016856766.1

View Full Product Details