Product Details

SNP ID
rs11998667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116962705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCACGAGGTAATATTGTATAACC[A/G]GAGTCATGTCTAAAGGTGCTGCTTT
Phenotype
MIM: 611145
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A8 PubMed Links

Gene Details

Gene
SLC30A8
Gene Name
solute carrier family 30 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172811.1 Intron NP_001166282.1
NM_001172813.1 Intron NP_001166284.1
NM_001172814.1 Intron NP_001166285.1
NM_001172815.2 Intron NP_001166286.1
NM_173851.2 Intron NP_776250.2

View Full Product Details