Product Details
- SNP ID
-
rs2034310
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74695789 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAGGAAAGCTTCGTGGTAGCCTTT[C/T]GCGGGGAGGTTCCGGCCAGCTGCAG
- Phenotype
-
MIM: 609807
MIM: 609956
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD300LF
PubMed Links
Gene Details
- Gene
- CD300LF
- Gene Name
- CD300 molecule like family member f
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289082.1 |
863 |
Silent Mutation |
GCA,GCG |
A183A |
NP_001276011.1 |
NM_001289083.1 |
863 |
Silent Mutation |
GCA,GCG |
A233A |
NP_001276012.1 |
NM_001289084.1 |
863 |
Missense Mutation |
CAA,CGA |
Q233R |
NP_001276013.1 |
NM_001289085.1 |
863 |
Missense Mutation |
CAA,CGA |
Q221R |
NP_001276014.1 |
NM_001289086.1 |
863 |
Silent Mutation |
GCA,GCG |
A228A |
NP_001276015.1 |
NM_001289087.1 |
863 |
Silent Mutation |
GCA,GCG |
A210A |
NP_001276016.1 |
NM_139018.4 |
863 |
Missense Mutation |
CAA,CGA |
Q218R |
NP_620587.2 |
XM_017024212.1 |
863 |
Missense Mutation |
CAA,CGA |
Q237R |
XP_016879701.1 |
XM_017024213.1 |
863 |
Silent Mutation |
GCA,GCG |
A255A |
XP_016879702.1 |
XM_017024214.1 |
863 |
Silent Mutation |
GCA,GCG |
A252A |
XP_016879703.1 |
XM_017024215.1 |
863 |
Silent Mutation |
GCA,GCG |
A236A |
XP_016879704.1 |
XM_017024216.1 |
863 |
Silent Mutation |
GCA,GCG |
A229A |
XP_016879705.1 |
XM_017024217.1 |
863 |
Silent Mutation |
GCA,GCG |
A225A |
XP_016879706.1 |
- Gene
- RAB37
- Gene Name
- RAB37, member RAS oncogene family
View Full Product Details