Product Details

SNP ID
rs1061115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98417292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCATCTCCCAGGATAAAGGCAGAG[A/G]CAGCTCTGGAAGAGGAGCTGTTGCC
Phenotype
MIM: 604982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPS1 PubMed Links

Gene Details

Gene
HPS1
Gene Name
HPS1, biogenesis of lysosomal organelles complex 3 subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000195.4 2566 UTR 3 NP_000186.2
NM_001311345.1 2566 UTR 3 NP_001298274.1
NM_001322476.1 2566 UTR 3 NP_001309405.1
NM_001322477.1 2566 UTR 3 NP_001309406.1
NM_001322478.1 2566 UTR 3 NP_001309407.1
NM_001322479.1 2566 UTR 3 NP_001309408.1
NM_001322480.1 2566 UTR 3 NP_001309409.1
NM_001322481.1 2566 UTR 3 NP_001309410.1
NM_001322482.1 2566 UTR 3 NP_001309411.1
NM_001322483.1 2566 UTR 3 NP_001309412.1
NM_001322484.1 2566 UTR 3 NP_001309413.1
NM_001322485.1 2566 UTR 3 NP_001309414.1
NM_001322487.1 2566 UTR 3 NP_001309416.1
NM_001322489.1 2566 UTR 3 NP_001309418.1
NM_001322490.1 2566 Intron NP_001309419.1
NM_001322491.1 2566 Intron NP_001309420.1
NM_001322492.1 2566 Intron NP_001309421.1
NM_182639.3 2566 Intron NP_872577.1
XM_005269757.4 2566 UTR 3 XP_005269814.1
XM_017016170.1 2566 UTR 3 XP_016871659.1
XM_017016171.1 2566 UTR 3 XP_016871660.1
XM_017016172.1 2566 UTR 3 XP_016871661.1
XM_017016173.1 2566 Intron XP_016871662.1
Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
There are no transcripts associated with this gene.

View Full Product Details