Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001288715.1 | 3986 | UTR 3 | NP_001275644.1 | ||
NM_001288716.1 | 3986 | UTR 3 | NP_001275645.1 | ||
NM_001288717.1 | 3986 | UTR 3 | NP_001275646.1 | ||
NM_001332.3 | 3986 | UTR 3 | NP_001323.1 | ||
XM_005248251.3 | 3986 | UTR 3 | XP_005248308.1 | ||
XM_005248252.2 | 3986 | UTR 3 | XP_005248309.1 | ||
XM_005248253.1 | 3986 | UTR 3 | XP_005248310.1 | ||
XM_011513967.2 | 3986 | UTR 3 | XP_011512269.1 | ||
XM_017009072.1 | 3986 | UTR 3 | XP_016864561.1 | ||
XM_017009073.1 | 3986 | UTR 3 | XP_016864562.1 | ||
XM_017009074.1 | 3986 | UTR 3 | XP_016864563.1 | ||
XM_017009075.1 | 3986 | UTR 3 | XP_016864564.1 | ||
XM_017009076.1 | 3986 | UTR 3 | XP_016864565.1 |