Product Details

SNP ID
rs3764880
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:12906707 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGAAAAATTAGAACAACAGAAAC[A/G]TGGTAAGCCACTTCTATTTCTTTAG
Phenotype
MIM: 300366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR8 PubMed Links

Gene Details

Gene
TLR8
Gene Name
toll like receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016610.3 88 UTR 5 NP_057694.2
NM_138636.5 88 Missense Mutation ATG,GTG M1V NP_619542.1
XM_011545529.1 88 UTR 5 XP_011543831.1
XM_011545530.2 88 UTR 5 XP_011543832.1
Gene
TLR8-AS1
Gene Name
TLR8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details