Product Details

SNP ID
rs1007923
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:6904943 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTCTCAACGGTGGCGAGCTGCAGT[A/T]GCCAAGTGCCCCGTCCCCGTTGCCA
Phenotype
MIM: 131360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENO2 PubMed Links

Gene Details

Gene
ENO2
Gene Name
enolase 2
There are no transcripts associated with this gene.

Gene
LRRC23
Gene Name
leucine rich repeat containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135217.1 211 UTR 5 NP_001128689.1
NM_006992.3 211 Intron NP_008923.1
NM_201650.2 211 Intron NP_964013.1

View Full Product Details