Product Details
- SNP ID
-
rs12618524
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:98447566 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCTAATTTTTTTTTTTTAGTATT[A/T]TGATATAGTTTCAGACTTACAGAAA
- Phenotype
-
MIM: 600916
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
INPP4A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76111899] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INPP4A
- Gene Name
- inositol polyphosphate-4-phosphatase type I A
View Full Product Details