Product Details

SNP ID
rs9346641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:168009397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTAGTGATTTGCATATTTTGAAC[A/T]GTCCTCACATTTTGGGATAAATCCC
Phenotype
MIM: 603815
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF25 PubMed Links

Gene Details

Gene
KIF25
Gene Name
kinesin family member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005355.3 Intron NP_005346.3
NM_030615.2 Intron NP_085118.2
XM_011535802.2 Intron XP_011534104.1
XM_011535803.2 Intron XP_011534105.1

View Full Product Details