Product Details
- SNP ID
-
rs11861531
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3066324 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGTTTCCATGATGTGGCCTGGC[C/T]CAGGTTCAGGACAGTGACCCGGAGG
- Phenotype
-
MIM: 606001
MIM: 608482
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL32
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115567031] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL32
- Gene Name
- interleukin 32
- Gene
- MMP25
- Gene Name
- matrix metallopeptidase 25
There are no transcripts associated with this gene.
- Gene
- MMP25-AS1
- Gene Name
- MMP25 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details