Product Details
- SNP ID
-
rs362012
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:22516821 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAGAACTTTTGTTGTTCCCGGAC[A/G]TTGCGTGCCTCTTTCCCCAGATTAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF280A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114413512,rs116793938] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF280A
- Gene Name
- zinc finger protein 280A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080740.4 |
|
Intron |
|
|
NP_542778.2 |
- Gene
- ZNF280B
- Gene Name
- zinc finger protein 280B
There are no transcripts associated with this gene.
View Full Product Details