Product Details

SNP ID
rs213205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33273358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTCCACAAACCTAATGAATTC[C/T]TGGCTTTCTAGATACATAACGTTCT
Phenotype
MIM: 603095 MIM: 180473 MIM: 603443 MIM: 611440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALT4 PubMed Links

Gene Details

Gene
B3GALT4
Gene Name
beta-1,3-galactosyltransferase 4
There are no transcripts associated with this gene.

Gene
RPS18
Gene Name
ribosomal protein S18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022551.2 Intron NP_072045.1
Gene
VPS52
Gene Name
VPS52, GARP complex subunit
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details