Product Details
- SNP ID
-
rs140392
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:21108064 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTATAATTATAACATGTCAGCAAGA[A/G]AACCAAAAATCACCTTTCAAATCTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCRP2
PubMed Links
Gene Details
- Gene
- BCRP2
- Gene Name
- breakpoint cluster region pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details