Product Details

SNP ID
rs564627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117290042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTAATAGGCTTAAAGAAATCAG[G/T]CTAAAGGAAAGTTTAGAATTTCAAC
Phenotype
MIM: 165020
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ROS1 PubMed Links

Gene Details

Gene
ROS1
Gene Name
ROS proto-oncogene 1, receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002944.2 Intron NP_002935.2
XM_006715548.3 Intron XP_006715611.1
XM_011536049.2 Intron XP_011534351.1
XM_011536050.2 Intron XP_011534352.1
XM_011536051.2 Intron XP_011534353.1
XM_011536052.2 Intron XP_011534354.1
XM_011536053.2 Intron XP_011534355.1
XM_011536054.2 Intron XP_011534356.1
XM_011536055.2 Intron XP_011534357.1
XM_011536056.2 Intron XP_011534358.1
XM_011536057.2 Intron XP_011534359.1
XM_011536058.2 Intron XP_011534360.1
XM_017011172.1 Intron XP_016866661.1
XM_017011173.1 Intron XP_016866662.1

View Full Product Details