Product Details

SNP ID
rs132567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39577521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGTTGAGGCTGTCGAAGGAGGG[A/G]ATGGCTCGTGTTCATCCATTGTTCT
Phenotype
MIM: 608230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA1I PubMed Links

Gene Details

Gene
CACNA1I
Gene Name
calcium voltage-gated channel subunit alpha1 I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003406.1 Intron NP_001003406.1
NM_021096.3 Intron NP_066919.2
XM_017029035.1 Intron XP_016884524.1
XM_017029036.1 Intron XP_016884525.1
XM_017029037.1 Intron XP_016884526.1

View Full Product Details