Product Details
- SNP ID
-
rs139859
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37945127 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTTCCGTTCCTCCATGTCCTTCCC[C/T]GTGGCAAAGATATTTTGGAGTCTTT
- Phenotype
-
MIM: 604414
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C22orf23
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35562630] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C22orf23
- Gene Name
- chromosome 22 open reading frame 23
- Gene
- MICALL1
- Gene Name
- MICAL like 1
There are no transcripts associated with this gene.
- Gene
- POLR2F
- Gene Name
- RNA polymerase II subunit F
There are no transcripts associated with this gene.
View Full Product Details