Product Details

SNP ID
rs623360
Assay Type
Validated
NCBI dbSNP Submissions
41
Location
Chr.1:209779520 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GGGATGTGTGGGAGCTTTTTAAGGA[C/G]TGATCATTGGCTCTGAGGACACTTC
Phenotype
MIM: 607199 MIM: 608255
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf74 PubMed Links

Gene Details

Gene
C1orf74
Gene Name
chromosome 1 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152485.3 1763 UTR 3 NP_689698.1
Gene
IRF6
Gene Name
interferon regulatory factor 6
There are no transcripts associated with this gene.

Gene
TRAF3IP3
Gene Name
TRAF3 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287754.1 1763 UTR 3 NP_001274683.1
NM_001320143.1 1763 Intron NP_001307072.1
NM_001320144.1 1763 Intron NP_001307073.1
NM_025228.3 1763 Intron NP_079504.2
XM_005273279.4 1763 Intron XP_005273336.1
XM_005273280.2 1763 Intron XP_005273337.1
XM_011510018.2 1763 UTR 3 XP_011508320.1
XM_011510019.1 1763 Intron XP_011508321.1
XM_017002399.1 1763 Intron XP_016857888.1
XM_017002400.1 1763 Intron XP_016857889.1

View Full Product Details