Product Details

SNP ID
rs563347
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:9297901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGATCTTTCACGCATACGTTGAG[A/G]AATAGATTTGTGAGGGGGCCCCAGC
Phenotype
MIM: 611657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPSB1 PubMed Links

Gene Details

Gene
SPSB1
Gene Name
splA/ryanodine receptor domain and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025106.3 Intron NP_079382.2

View Full Product Details