Product Details

SNP ID
rs656150
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109376743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTACTTGGTTGTCAAGAACCACA[A/G]GTCTCAGAAGAGAAAAGGAGTGTAA
Phenotype
MIM: 300041
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GUCY2F PubMed Links

Gene Details

Gene
GUCY2F
Gene Name
guanylate cyclase 2F, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001522.2 Intron NP_001513.2

View Full Product Details