Product Details
- SNP ID
-
hCV2247157
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:26185332 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CATGAATTACCAGAAAGACCACCCA[A/G]TGAGAGTCAGAGACCTGAGTTCTGG
- Phenotype
-
MIM: 607021
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEZ6L
PubMed Links
Gene Details
- Gene
- SEZ6L
- Gene Name
- seizure related 6 homolog like
There are no transcripts associated with this gene.
View Full Product Details