Product Details

SNP ID
rs223074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32096273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGGAGGTGTTTTTGAAGTTCCC[C/T]ACTGGGATGCTGTCCTGGTTTGGGA
Phenotype
MIM: 602735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RCN1 PubMed Links

Gene Details

Gene
RCN1
Gene Name
reticulocalbin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002901.2 Intron NP_002892.1

View Full Product Details