Product Details

SNP ID
rs418485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75256553 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGTAAAGGTTATGTAGTTTCTC[A/G]GGTGCTTTTGCTCTCATTACATGGA
Phenotype
MIM: 611028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM30A PubMed Links

Gene Details

Gene
TMEM30A
Gene Name
transmembrane protein 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143958.1 Intron NP_001137430.1
NM_018247.3 Intron NP_060717.1

View Full Product Details