Product Details

SNP ID
rs174699
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:19966935 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCTCTGACACTGTCGCTTCTCCA[T/C]GGCATTAGATTTTCAGTCCTGCTCA
Phenotype
MIM: 602269 MIM: 116790
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ARVCF PubMed Links

Gene Details

Gene
ARVCF
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001670.2 1954 Intron NP_001661.1
XM_005261242.2 1954 UTR 3 XP_005261299.1
XM_005261243.3 1954 Intron XP_005261300.1
XM_005261244.3 1954 Intron XP_005261301.1
XM_006724243.2 1954 UTR 3 XP_006724306.1
XM_006724245.2 1954 Intron XP_006724308.1
XM_006724246.3 1954 UTR 3 XP_006724309.1
XM_006724247.3 1954 Intron XP_006724310.1
XM_006724248.3 1954 Intron XP_006724311.1
XM_006724249.3 1954 Intron XP_006724312.1
XM_006724250.3 1954 Intron XP_006724313.1
XM_011530179.2 1954 UTR 3 XP_011528481.1
XM_011530180.1 1954 Intron XP_011528482.1
XM_011530181.1 1954 Intron XP_011528483.1
XM_011530182.2 1954 UTR 3 XP_011528484.1
XM_011530183.2 1954 Intron XP_011528485.1
Gene
COMT
Gene Name
catechol-O-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000754.3 1954 Intron NP_000745.1
NM_001135161.1 1954 Intron NP_001128633.1
NM_001135162.1 1954 Intron NP_001128634.1
NM_007310.2 1954 Intron NP_009294.1
XM_011529886.1 1954 Intron XP_011528188.1
XM_017028594.1 1954 Intron XP_016884083.1
XM_017028595.1 1954 Intron XP_016884084.1
Gene
MIR4761
Gene Name
microRNA 4761
There are no transcripts associated with this gene.

View Full Product Details