Product Details

SNP ID
rs730927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40924658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAGTCTCAAGGTCATGGGTTCA[C/T]GGGAGAACCCGTCTGGATACAGAAG
Phenotype
MIM: 606194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT23 PubMed Links

Gene Details

Gene
KRT23
Gene Name
keratin 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282433.1 Intron NP_001269362.1
NM_015515.4 Intron NP_056330.3
XM_005257200.4 Intron XP_005257257.1
XM_011524595.1 Intron XP_011522897.1

View Full Product Details