Product Details

SNP ID
rs756462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136032256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCTCCAGCCTTCACTTCTCTTG[C/T]CCCTAGATCCTTACATGGATTCATT
Phenotype
MIM: 601692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGFBI PubMed Links

Gene Details

Gene
TGFBI
Gene Name
transforming growth factor beta induced
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000358.2 Intron NP_000349.1

View Full Product Details