Product Details

SNP ID
rs178257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20968782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCAGCCAGTGCTCCACTCAACC[C/T]GCCAGGCCACGCTGCCCCAGCCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIFM3 PubMed Links

Gene Details

Gene
AIFM3
Gene Name
apoptosis inducing factor, mitochondria associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018060.2 Intron NP_001018070.1
NM_001146288.1 Intron NP_001139760.1
NM_144704.2 Intron NP_653305.1
Gene
LOC101928891
Gene Name
uncharacterized LOC101928891
There are no transcripts associated with this gene.

View Full Product Details