Product Details
- SNP ID
-
hCV2293049
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:25340112 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGTGTCTGGGCCATTTTTGGCT[A/T]TAATCATCTTTAATTTTCCTAGTCC
- Phenotype
-
MIM: 601623
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UBE3A
PubMed Links
Gene Details
- Gene
- UBE3A
- Gene Name
- ubiquitin protein ligase E3A
There are no transcripts associated with this gene.
View Full Product Details