Product Details

SNP ID
rs758853
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:16000288 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGCGCCCGGCCGAGCGCGGTAGC[A/C]TTTGTGAGCGGGGTCTTGGCGTTGC
Phenotype
MIM: 613814 MIM: 614535
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTC19 PubMed Links

Gene Details

Gene
TTC19
Gene Name
tetratricopeptide repeat domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271420.1 824 UTR 5 NP_001258349.1
NM_017775.3 824 Intron NP_060245.3
XM_017024801.1 824 Intron XP_016880290.1
XM_017024802.1 824 Intron XP_016880291.1
Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 824 Intron NP_001036162.1
NM_001042698.1 824 Intron NP_001036163.1

View Full Product Details