Product Details

SNP ID
rs26351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42363436 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTATCGTGATAGCTGCAATCTAT[C/T]TTCAGCAAAAATAAAATATATACAC
Phenotype
MIM: 612750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYZL4 PubMed Links

Gene Details

Gene
LYZL4
Gene Name
lysozyme like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304386.1 Intron NP_001291315.1
NM_144634.3 Intron NP_653235.1
XM_011533355.2 Intron XP_011531657.1
XM_017005705.1 Intron XP_016861194.1
XM_017005706.1 Intron XP_016861195.1

View Full Product Details