Product Details
- SNP ID
-
rs626783
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3709741 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTGAGATTCGGGAGTCCGTGGGG[G/T]TGGGCGACATGGATCCCACCTTGCC
- Phenotype
-
MIM: 606102
MIM: 612689
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PIP5K1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76659614] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PIP5K1C
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
There are no transcripts associated with this gene.
- Gene
- TJP3
- Gene Name
- tight junction protein 3
View Full Product Details