Product Details

SNP ID
rs333075
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:110032239 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTTAGCTGAAAAATGTTTTTCCCC[A/G]GGTGATACAGCTTATGATAGGCACA
Phenotype
MIM: 607826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHCYL1 PubMed Links

Gene Details

Gene
AHCYL1
Gene Name
adenosylhomocysteinase like 1
There are no transcripts associated with this gene.

Gene
STRIP1
Gene Name
striatin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270768.1 Intron NP_001257697.1
NM_033088.3 Intron NP_149079.2
XM_006710995.2 Intron XP_006711058.1
XM_017002629.1 Intron XP_016858118.1
XM_017002630.1 Intron XP_016858119.1

View Full Product Details