Product Details

SNP ID
rs605367
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:23192031 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ATGGTGTCCCACTCAAAGCTTGCGG[C/T]GGGTGGGGGGTGGGCGTCTAAAGCC
Phenotype
MIM: 182133
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR1D PubMed Links

Gene Details

Gene
HTR1D
Gene Name
5-hydroxytryptamine receptor 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000864.4 2699 UTR 3 NP_000855.1

View Full Product Details