Product Details

SNP ID
rs624270
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:210233595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCTCCTCCCAGGCAGCAGAGC[C/T]GCGCTGCGGGCGGCGGCGGCTCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERTAD4 PubMed Links

Gene Details

Gene
SERTAD4
Gene Name
SERTA domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019605.3 Intron NP_062551.1
XM_005273193.4 Intron XP_005273250.1
Gene
SERTAD4-AS1
Gene Name
SERTAD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details