Product Details

SNP ID
rs724849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30601206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTATGGCTGTGGTCTTAGCGGCT[A/G]TGGCTATGCCTGCTACTGCCCATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP22-1 PubMed Links

Gene Details

Gene
KRTAP22-1
Gene Name
keratin associated protein 22-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181620.1 86 Missense Mutation TAT,TGT Y29C NP_853651.1
Gene
KRTAP6-2
Gene Name
keratin associated protein 6-2
There are no transcripts associated with this gene.

Gene
KRTAP6-3
Gene Name
keratin associated protein 6-3
There are no transcripts associated with this gene.

View Full Product Details