Product Details

SNP ID
rs3887397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140692298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCATAGTATGGAACCACCACAT[C/T]CTCTATTTTTGGAAGGGCATACAGC
Phenotype
MIM: 142810 MIM: 600783
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HARS PubMed Links

Gene Details

Gene
HARS
Gene Name
histidyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258040.2 Intron NP_001244969.1
NM_001258041.2 Intron NP_001244970.1
NM_001258042.2 Intron NP_001244971.1
NM_001289092.1 Intron NP_001276021.1
NM_001289093.1 Intron NP_001276022.1
NM_001289094.1 Intron NP_001276023.1
NM_002109.5 Intron NP_002100.2
Gene
HARS2
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278731.1 Intron NP_001265660.1
NM_001278732.1 Intron NP_001265661.1
NM_012208.3 Intron NP_036340.1
Gene
ZMAT2
Gene Name
zinc finger matrin-type 2
There are no transcripts associated with this gene.

View Full Product Details