Product Details

SNP ID
rs39794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40834331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACACCTTGTAGGTGTTGTTAA[C/T]ACACGAGTTTTATGCCACCTCATCG
Phenotype
MIM: 609986 MIM: 604181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD6 PubMed Links

Gene Details

Gene
CARD6
Gene Name
caspase recruitment domain family member 6
There are no transcripts associated with this gene.

Gene
LOC100506548
Gene Name
uncharacterized LOC100506548
There are no transcripts associated with this gene.

Gene
RPL37
Gene Name
ribosomal protein L37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000997.4 Intron NP_000988.1
Gene
SNORD72
Gene Name
small nucleolar RNA, C/D box 72
There are no transcripts associated with this gene.

View Full Product Details