Product Details

SNP ID
rs190414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56820945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTCTGATAAGAATATATACAA[A/C]ACTGCATGTTCAGGCATGTTTTTTT
Phenotype
MIM: 600982
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K1 PubMed Links

Gene Details

Gene
MAP3K1
Gene Name
mitogen-activated protein kinase kinase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005921.1 Intron NP_005912.1
XM_017009484.1 Intron XP_016864973.1
XM_017009485.1 Intron XP_016864974.1

View Full Product Details