Product Details

SNP ID
rs245114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135571451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCCAAGCTAGGAATGGAGCAA[C/T]ACTGCAATGAAATGTGTCCACCAAG
Phenotype
MIM: 604186 MIM: 616150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL14 PubMed Links

Gene Details

Gene
CXCL14
Gene Name
C-X-C motif chemokine ligand 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004887.4 1203 UTR 3 NP_004878.2
Gene
SLC25A48
Gene Name
solute carrier family 25 member 48
There are no transcripts associated with this gene.

View Full Product Details