Product Details

SNP ID
rs763666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72044447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGAACTCTTTGAAGGCAGAGG[A/C]TATGTTTATCCCCCATTCAATTCAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNL4B PubMed Links

Gene Details

Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 Intron NP_001135789.1
NM_001142318.1 Intron NP_001135790.1
NM_001324354.1 Intron NP_001311283.1
NM_001324355.1 Intron NP_001311284.1
NM_017853.2 Intron NP_060323.1

View Full Product Details