Product Details
- SNP ID
-
hCV2394180
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71568278 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCACTCGGATGAAATTCGGGTACT[A/C]AAAGCACTGCAAAAAGAAGAGTCTG
- Phenotype
-
MIM: 613018
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TAT
PubMed Links
Gene Details
- Gene
- TAT
- Gene Name
- tyrosine aminotransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000353.2 |
1330 |
Nonsense Mutation |
GAG,TAG |
E411* |
NP_000344.1 |
- Gene
- TAT-AS1
- Gene Name
- TAT antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details