Product Details

SNP ID
rs399209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84295960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACTCAATGGGGGGCTGGGGGCAG[C/T]AAAGAATAAATGAGAGAGATCTCAT
Phenotype
MIM: 605322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376777 PubMed Links

Gene Details

Gene
LOC105376777
Gene Name
uncharacterized LOC105376777
There are no transcripts associated with this gene.

Gene
WFDC1
Gene Name
WAP four-disulfide core domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282466.1 Intron NP_001269395.1
NM_001282467.1 Intron NP_001269396.1
NM_021197.3 Intron NP_067020.2
XM_005256079.3 Intron XP_005256136.1

View Full Product Details