Product Details

SNP ID
rs39550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84221240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGCCAGCAAGGGACAGCACCTA[C/T]AATTCCCCTTGTTTCTGTGCCAAAG
Phenotype
MIM: 607603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNG4 PubMed Links

Gene Details

Gene
KCNG4
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172347.2 2658 UTR 3 NP_758857.1

View Full Product Details